ENA371463168 Enamine

1-(furan-2-yl)-2-(morpholin-3-yl)ethan-1-one hydrochloride

  • MDL number MFCD22378642
  • Cas Number : 1394041-99-6
  • Molecular Weight : 231.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-furyl)-2-(3-morpholinyl)ethanone hydrochloride
Inchi Code 1S/C10H13NO3.ClH/c12-9(10-2-1-4-14-10)6-8-7-13-5-3-11-8;/h1-2,4,8,11H,3,5-7H2;1H
Inchi Key XVNZGZXALXHRMQ-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-108935

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108935
Availability details for
Quantity
  • Availability
  • Add to Cart