ENA371463173 Enamine

methyl 2-{6-chloro-3-oxo-2H,3H-[1,2,4]triazolo[4,3-b]pyridazin-2-yl}acetate

  • MDL number MFCD22378647
  • Cas Number : 1394041-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108954

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108954
Availability details for
Quantity
  • Availability
  • Add to Cart