ENA371463177 Enamine

2-chloro-N-[2-(quinolin-8-yl)ethyl]acetamide hydrochloride

  • MDL number MFCD22378651
  • Cas Number : 1394042-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186...188
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109043

Safety Information

msds https://www.enaminestore.com/catalog/EN300-109043
Availability details for
Quantity
  • Availability
  • Add to Cart