ENA371463178 Enamine

1-(pyridine-2-carbonyl)pyrrolidin-3-amine dihydrochloride

  • MDL number MFCD22378652
  • Cas Number : 1394040-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109056

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109056
Availability details for
Quantity
  • Availability
  • Add to Cart