ENA371463180 Enamine

3-oxo-3-(piperazin-1-yl)propanenitrile hydrochloride

  • MDL number MFCD22378654
  • Cas Number : 1394042-17-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109060

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109060
Availability details for
Quantity
  • Availability
  • Add to Cart