ENA371463182 Enamine

2-[4-(aminomethyl)-1H-1,2,3-triazol-1-yl]acetic acid hydrochloride

  • MDL number MFCD22378656
  • Cas Number : 1394041-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109069

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109069
Availability details for
Quantity
  • Availability
  • Add to Cart