ENA371463186 Enamine

N-methoxy-2,3,4,5-tetrahydro-1-benzoxepin-5-amine

  • MDL number MFCD22378661
  • Cas Number : 1394042-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109126

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109126
Availability details for
Quantity
  • Availability
  • Add to Cart