ENA371463187 Enamine

5-methyl-2,3-dihydro-1-benzofuran-3-amine hydrochloride

  • MDL number MFCD22378662
  • Cas Number : 669-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 339...341
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109136

Safety Information

msds https://www.enaminestore.com/catalog/EN300-109136
Availability details for
Quantity
  • Availability
  • Add to Cart