ENA371463189 Enamine

3-(4-tert-butyl-2-methylphenoxy)propan-1-amine hydrochloride

  • MDL number MFCD22378664
  • Cas Number : 1394042-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109138

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109138
Availability details for
Quantity
  • Availability
  • Add to Cart