ENA371463197 Enamine

ethyl 1-(2-cyanoethyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD22378674
  • Cas Number : 1394041-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109369

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109369
Availability details for
Quantity
  • Availability
  • Add to Cart