ENA371463199 Enamine

N-(1,2,3,4-tetrahydroquinolin-5-yl)butanamide hydrochloride

  • MDL number MFCD22378676
  • Cas Number : 1394040-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109391

Safety Information

msds https://www.enaminestore.com/catalog/EN300-109391
Availability details for
Quantity
  • Availability
  • Add to Cart