ENA371463200 Enamine

3,5-dimethyl-4-sulfamoyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD22378677
  • Cas Number : 1394041-61-2
  • Molecular Weight : 218.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-109393

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109393
Availability details for
Quantity
  • Availability
  • Add to Cart