ENA371463202 Enamine

3-(difluoromethyl)-5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyridine-6-carboxylic acid

  • MDL number MFCD22378679
  • Cas Number : 1394042-15-9
  • Molecular Weight : 217.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 216-219
Iupac Name 3-(difluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyridine-6-carboxylic acid
Inchi Code 1S/C8H9F2N3O2/c9-6(10)7-12-11-5-2-1-4(8(14)15)3-13(5)7/h4,6H,1-3H2,(H,14,15)
Inchi Key NKZZIJAHOJMTFA-UHFFFAOYSA-N

Description

Melting Point 216-219
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-109481

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-109481
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart