ENA371463211 Enamine

sodium (1-oxo-2,3-dihydro-1H-inden-2-ylidene)methanolate

  • MDL number MFCD18870123
  • Cas Number : 1344812-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109591

Safety Information

msds https://www.enaminestore.com/catalog/EN300-109591
Availability details for
Quantity
  • Availability
  • Add to Cart