ENA371463221 Enamine

2-(2-chlorophenyl)-2-methoxyethan-1-amine hydrochloride

  • MDL number MFCD22378711
  • Cas Number : 1394040-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109807

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109807
Availability details for
Quantity
  • Availability
  • Add to Cart