ENA371463226 Enamine

5-(chlorosulfonyl)-2-fluoro-3-methylbenzoic acid

  • MDL number MFCD22378716
  • Cas Number : 1394040-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-109859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109859
Availability details for
Quantity
  • Availability
  • Add to Cart