ENA371463230 Enamine

2-amino-5-oxohexanoic acid hydrochloride

  • MDL number MFCD22378720
  • Cas Number : 1394040-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109927

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109927
Availability details for
Quantity
  • Availability
  • Add to Cart