ENA371463237 Enamine

1-(2-aminoethoxy)-3,5-dimethoxybenzene hydrochloride

  • MDL number MFCD22378727
  • Cas Number : 61711-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110006

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110006
Availability details for
Quantity
  • Availability
  • Add to Cart