ENA371463242 Enamine

4-(dimethylamino)-3-fluorobenzene-1-carboximidamide dihydrochloride

  • MDL number MFCD22378733
  • Cas Number : 1394042-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110038

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110038
Availability details for
Quantity
  • Availability
  • Add to Cart