ENA371463247 Enamine

1-[3-(dimethylamino)propyl]imidazolidine-2,4,5-trione hydrochloride

  • MDL number MFCD22378738
  • Cas Number : 1394042-21-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-110106

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110106
Availability details for
Quantity
  • Availability
  • Add to Cart