ENA371463264 Enamine

methyl N-(1,2,3,4-tetrahydroisoquinolin-5-yl)carbamate hydrochloride

  • MDL number MFCD22378756
  • Cas Number : 1394041-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-110335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110335
Availability details for
Quantity
  • Availability
  • Add to Cart