ENA371463274 Enamine

3-(hydrazinylmethyl)quinoline dihydrochloride

  • MDL number MFCD22378765
  • Cas Number : 1365836-82-3
  • Molecular Weight : 246.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(hydrazinomethyl)quinoline dihydrochloride
Inchi Code 1S/C10H11N3.2ClH/c11-13-7-8-5-9-3-1-2-4-10(9)12-6-8;;/h1-6,13H,7,11H2;2*1H
Inchi Key LINQNBNLOBJGKN-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-110502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110502
Availability details for
Quantity
  • Availability
  • Add to Cart