ENA371463282 Enamine

sodium 2-chlorobenzene-1-sulfinate

  • MDL number MFCD11618026
  • Cas Number : 15946-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-105395

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105395
Availability details for
Quantity
  • Availability
  • Add to Cart