ENA371463288 Enamine

2-(4-cyanobenzenesulfonamido)propanoic acid

  • MDL number MFCD11121733
  • Cas Number : 1308963-00-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108673

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108673
Availability details for
Quantity
  • Availability
  • Add to Cart