ENA371463292 Enamine

1-(2,2,2-trifluoroethyl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD08696335
  • Cas Number : 790254-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105517

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105517
Availability details for
Quantity
  • Availability
  • Add to Cart