ENA371463295 Enamine

2,4-di-tert-butyl-6-chloropyrimidine

  • MDL number MFCD00234810
  • Cas Number : 69050-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108682

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108682
Availability details for
Quantity
  • Availability
  • Add to Cart