ENA371463296 Enamine

5-(chloromethyl)-3-(4-fluoro-3-methylphenyl)-1,2,4-oxadiazole

  • MDL number MFCD09051121
  • Cas Number : 937629-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108811

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108811
Availability details for
Quantity
  • Availability
  • Add to Cart