ENA371463301 Enamine

2-bromo-5-(difluoromethyl)-1,3,4-thiadiazole

  • MDL number MFCD18345064
  • Cas Number : 1340313-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108825

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108825
Availability details for
Quantity
  • Availability
  • Add to Cart