ENA371463302 Enamine

3,5-dinitro-1,4-dihydropyridin-4-one

  • MDL number MFCD00179579
  • Cas Number : 19872-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-105697

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105697
Availability details for
Quantity
  • Availability
  • Add to Cart