ENA371463303 Enamine

(1R)-2,2,2-trifluoro-1-(4-methyl-1,3-thiazol-2-yl)ethan-1-ol

  • MDL number MFCD22378630
  • Cas Number : 1394051-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108827

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108827
Availability details for
Quantity
  • Availability
  • Add to Cart