ENA371463314 Enamine

5-bromo-4-hydroxy-6-methylpyridine-3-carboxylic acid

  • MDL number MFCD11557201
  • Cas Number : 141703-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108838

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108838
Availability details for
Quantity
  • Availability
  • Add to Cart