ENA371463315 Enamine

5-chloro-2,3-dihydro-1-benzofuran-3-amine hydrochloride

  • MDL number MFCD08544175
  • Cas Number : 669-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108839

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108839
Availability details for
Quantity
  • Availability
  • Add to Cart