ENA371463317 Enamine

4,6-di-tert-butylpyrimidin-2-amine

  • MDL number MFCD14588414
  • Cas Number : 78641-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108841

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108841
Availability details for
Quantity
  • Availability
  • Add to Cart