ENA371463322 Enamine

3-bromoquinoline-2-carbonitrile

  • MDL number MFCD08705682
  • Cas Number : 61830-11-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128-130
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108847

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108847
Availability details for
Quantity
  • Availability
  • Add to Cart