ENA371463324 Enamine

2-[3-(aminomethyl)oxolan-3-yl]ethan-1-ol

  • MDL number MFCD21689075
  • Cas Number : 1394040-43-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105829

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105829
Availability details for
Quantity
  • Availability
  • Add to Cart