ENA371463330 Enamine

spiro[2.5]octane-1-carboxylic acid

  • MDL number MFCD09864204
  • Cas Number : 17202-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105932

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105932
Availability details for
Quantity
  • Availability
  • Add to Cart