ENA371463342 Enamine

4-chloro-6-methylpyridine-3-carboxylic acid

  • MDL number MFCD11520850
  • Cas Number : 1060805-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106073

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-106073
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart