ENA371463343 Enamine

1-(5-bromo-3-chloro-2-hydroxyphenyl)ethan-1-one

  • MDL number MFCD02660600
  • Cas Number : 331821-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106332

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106332
Availability details for
Quantity
  • Availability
  • Add to Cart