ENA371463345 Enamine

4-[ethyl(methyl)amino]benzene-1-carboximidamide

  • MDL number MFCD11623025
  • Cas Number : 1094944-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108937
Availability details for
Quantity
  • Availability
  • Add to Cart