ENA371463348 Enamine

4-[ethyl(methyl)amino]-N'-hydroxybenzene-1-carboximidamide

  • MDL number MFCD11622983
  • Cas Number : 1094777-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108940

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108940
Availability details for
Quantity
  • Availability
  • Add to Cart