ENA371463350 Enamine

2-chloro-4-fluoro-5-methylbenzene-1-sulfonamide

  • MDL number MFCD12026247
  • Cas Number : 1208077-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108944

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108944
Availability details for
Quantity
  • Availability
  • Add to Cart