ENA371463356 Enamine

(4-methylpyridin-3-yl)methanol

  • MDL number MFCD01444898
  • Cas Number : 4664-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108952

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108952
Availability details for
Quantity
  • Availability
  • Add to Cart