ENA371463371 Enamine

6-bromo-5-methyl-3H-imidazo[4,5-b]pyridine-2-thiol

  • MDL number MFCD00186964
  • Cas Number : 104685-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109007

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109007
Availability details for
Quantity
  • Availability
  • Add to Cart