ENA371463377 Enamine

4-chloro-N1-(cyclohexylmethyl)benzene-1,2-diamine

  • MDL number MFCD12166789
  • Cas Number : 1096873-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109036

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109036
Availability details for
Quantity
  • Availability
  • Add to Cart