ENA371463384 Enamine

2,3-dichloro-1-benzothiophene

  • MDL number MFCD01681796
  • Cas Number : 5323-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53...55
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109051

Safety Information

msds https://www.enaminestore.com/catalog/EN300-109051
Availability details for
Quantity
  • Availability
  • Add to Cart