ENA371463386 Enamine

3-fluoro-4-[(4-iodophenoxy)methyl]benzamide

  • MDL number MFCD16386198
  • Cas Number : 1284147-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-109054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109054
Availability details for
Quantity
  • Availability
  • Add to Cart