ENA371463389 Enamine

5-methyloxolane-2-carboxylic acid

  • MDL number MFCD19228944
  • Cas Number : 61834-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-107103

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107103
Availability details for
Quantity
  • Availability
  • Add to Cart