ENA371463394 Enamine

1-(1-phenyl-1H-pyrazol-4-yl)ethan-1-ol

  • MDL number MFCD12787773
  • Cas Number : 104216-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109065

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109065
Availability details for
Quantity
  • Availability
  • Add to Cart