ENA371463398 Enamine

1-bromo-3-(1-bromoethyl)benzene

  • MDL number MFCD11180269
  • Cas Number : 59770-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103419

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103419
Availability details for
Quantity
  • Availability
  • Add to Cart