ENA371463399 Enamine

ethyl 3-cyclopropyl-2,2-difluoropropanoate

  • MDL number MFCD21363155
  • Cas Number : 1267593-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103572

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-103572
pictograms GHS02,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart